Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α,α,2,3-Pentafluorotoluene
α,α,α,2,3-Pentafluorotoluene
α,α,α,2,3-Pentafluorotoluene
Flammable
Irritant
α,α,α,2,3-Pentafluorotoluene
Supplier:
Apollo Scientific
Certificates
Expand image
Flammable
Irritant
α,α,α,2,3-Pentafluorotoluene
Supplier:
Apollo Scientific
About this item
CAS Number:
64248-59-5
:
Formula:
C₇H₃F₅
MDL Number:
MFCD01631429
:
182.09 g/mol
UN:
1993
ADR:
3,III
:
GHS02
:
GHS07
Storage Temperature:
Ambient