
2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride
Catalog # APOSPC0219-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
CAS Number:143096-86-0
Formula:C₈H₃ClF₂O₃
MDL Number:MFCD01631474