Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoroanisole
2,3-Difluoroanisole
2,3-Difluoroanisole
:
Apollo Scientific
2,3-Difluoroanisole
:
Apollo Scientific
:
134364-69-5
:
:
C₇H₆F₂O
:
MFCD01631562
:
144.12 g/mol
:
1.24 g/cm³ (25 °C)
:
62 °C
:
GHS07
:
Ambient