Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5-fluorobenzamide
2,4-Dichloro-5-fluorobenzamide 97%
2,4-Dichloro-5-fluorobenzamide 97%
2,4-Dichloro-5-fluorobenzamide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichloro-5-fluorobenzamide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
135748-33-3
:
Formula:
C₇H₄Cl₂FNO
MDL Number:
MFCD01631478
:
208.02 g/mol
Melting Pt:
158…160 °C
Storage Temperature:
Ambient