
3,5-Difluoro-N-fmoc-L-phenylalanine
Catalog # APOSPC0201-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:423.4 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:205526-24-5
Formula:C₂₄H₁₉F₂NO₄
MDL Number:MFCD00797579