

(1,2-Dichlorotrifluoroethyl)cyclohexane
Catalog # APOSPC0140-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.08 g/mol
Boiling Pt:83…84 °C/15mmHg
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:219904-98-0
Formula:C₈H₁₁Cl₂F₃
MDL Number:MFCD00155754