
α-Chloro-2,4-difluorotoulene 97%
Catalog # APOSPC0049-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.57 g/mol
Boiling Pt:60 °C/9.75mmHg
Density:1.32 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
CAS Number:452-07-3
Formula:C₇H₅ClF₂
MDL Number:MFCD01090988