Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4',4''-Trifluorotrityl bromide
4,4',4''-Trifluorotrityl bromide
4,4',4''-Trifluorotrityl bromide
4,4',4''-Trifluorotrityl bromide
APOSPC0030-1G
:
Apollo Scientific
:
4,4',4''-Trifluorotrityl bromide
APOSPC0030-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
377.20 g/mol
:
Ambient
:
200004-38-2
:
C₁₉H₁₂BrF₃
:
MFCD00191586