Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-p-cresol
3-Chloro-p-cresol 98%
3-Chloro-p-cresol 98%
3-Chloro-p-cresol 98%
APOSOR9971-5G
:
Apollo Scientific
:
3-Chloro-p-cresol 98%
APOSOR9971-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
142.58 g/mol
:
228 °C
:
48…53 °C
:
210-439-1
:
GHS05
:
GHS07
:
Ambient
:
615-62-3
:
C₇H₇ClO
:
MFCD00060319