
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29668-44-8
:
Formula:C₉H₈O₃
MDL Number:MFCD00010092
:164.16 g/mol
Boiling Pt:104…106 °C/15mmHg
Melting Pt:52…53 °C
:GHS07
Storage Temperature:Ambient