Order Entry
Export
ContactUsLinkComponent
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Irritant
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Catalog # APOSOR9830-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Catalog # APOSOR9830-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.62 g/mol
Melting Pt:228…230 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:119448-28-1
Formula:C₈H₇ClN₄O
MDL Number:MFCD06797385