Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14098-44-3
:
Formula:C₁₄H₂₀O₅
MDL Number:MFCD00005945
:268.31 g/mol
Melting Pt:78…80 °C
Storage Temperature:Ambient