
2,6-Dichlorophenylhydrazinium chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50709-36-9
:
Formula:C₆H₇Cl₃N₂
MDL Number:MFCD00012930
:213.49 g/mol
Melting Pt:225 °C (dec.)
Storage Temperature:Ambient
EINECS:256-730-7
UN:2811
ADR:6.1,III
:GHS07