Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine 95%
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine 95%
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
918538-05-3
:
Formula:
C₆H₃Cl₂N₃
MDL Number:
MFCD11044885
:
188.0100 g/mol
Storage Temperature:
Ambient