Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2H-Pyrano[3,2-b]pyridin-4(3H)-one
2H-Pyrano[3,2-b]pyridin-4(3H)-one 98%
2H-Pyrano[3,2-b]pyridin-4(3H)-one 98%
2H-Pyrano[3,2-b]pyridin-4(3H)-one 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2H-Pyrano[3,2-b]pyridin-4(3H)-one 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
405174-48-3
:
Formula:
C₈H₇NO₂
MDL Number:
MFCD11035881
:
149.15 g/mol
Storage Temperature:
Ambient