Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-4-(Boc-amino)cyclohexanol
cis-4-(Boc-amino)cyclohexanol 97+%
cis-4-(Boc-amino)cyclohexanol 97+%
:
Apollo Scientific
cis-4-(Boc-amino)cyclohexanol 97+%
:
Apollo Scientific
:
167081-25-6
:
MFCD15071470
:
C₁₁H₂₁NO₃
:
MFCD03844614
:
215.293 g/mol
:
GHS07
:
Ambient