Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-1,3-dimethyl-1H-pyrazole
5-Bromo-1,3-dimethyl-1H-pyrazole 95%
5-Bromo-1,3-dimethyl-1H-pyrazole 95%
5-Bromo-1,3-dimethyl-1H-pyrazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Bromo-1,3-dimethyl-1H-pyrazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
5744-70-7
:
Formula:
C₅H₇BrN₂
MDL Number:
MFCD08448181
:
175.03 g/mol
Storage Temperature:
Ambient