Order Entry
Export
Orders LinkContactUsLinkComponent
5-Hydroxy-2-nitropyridine 95%

5-Hydroxy-2-nitropyridine 95%

New Product
Supplier:   Apollo Scientific
5-Hydroxy-2-nitropyridine 95%
Supplier:  Apollo Scientific
New Product
Certificates
5-Hydroxy-2-nitropyridine 95%

5 Options Available Below

Product Details & Documents
Synonyms:  6-nitropyridin-3-ol
  • CAS Number:15206-26-5
  • Formula:C5H4N2O3
  • MDL Number:MFCD03032441
  • Molecular Weight:140.0969 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 21,80 €
    21,80 €
    1 * 100 mg
       21,80 €

  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 43,50 €
    43,50 €
    1 * 250 mg
       43,50 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 102,00 €
    102,00 €
    1 * 1 g
       102,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 438,00 €
    438,00 €
    1 * 5 g
       438,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 190,00 €
    2 190,00 €
    1 * 25 g
       2 190,00 €

    Specifications:
    New Product
    New Product