Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dimethoxy-1H-indole-2-carboxylic acid
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
Irritant
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
88210-96-2
:
Formula:
C₁₁H₁₁NO₄
MDL Number:
MFCD00015459
Melting Pt:
210 °C (dec.)
:
221.21 g/mol
:
GHS07
Storage Temperature:
Ambient