
2,5-Dihydroxybenzoic acid, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:490-79-9
:
Formula:C₇H₆O₄
MDL Number:MFCD00002460
:154.12 g/mol
Melting Pt:204…208 °C
EINECS:207-718-5
:GHS07
Storage Temperature:Ambient