
2-(4-Chlorobutyl)-1,3-dioxolane 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118336-86-0
:
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00043136
:150.6 g/mol
Boiling Pt:56…58 °C
Density:1.109 g/cm³ (25 °C)
Flash Pt:85 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06