Order Entry
Export
ContactUsLinkComponent
1,2-Dimethylbenzimidazole 98%
1,2-Dimethylbenzimidazole 98%
Catalog # APOSOR954313-5G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Dimethylbenzimidazole 98%
Catalog # APOSOR954313-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.19 g/mol
Storage Temperature:Ambient
CAS Number:2876-08-6
Formula:C₉H₁₀N₂
MDL Number:MFCD00022840