Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dimethoxynaphthalene
1,5-Dimethoxynaphthalene 95%
1,5-Dimethoxynaphthalene 95%
1,5-Dimethoxynaphthalene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,5-Dimethoxynaphthalene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
10075-63-5
:
Formula:
C₁₂H₁₂O₂
MDL Number:
MFCD00086779
:
188.23 g/mol
Storage Temperature:
Ambient