Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromofuro[2,3-b]pyridine
5-Bromofuro[2,3-b]pyridine 98%
5-Bromofuro[2,3-b]pyridine 98%
5-Bromofuro[2,3-b]pyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Bromofuro[2,3-b]pyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
220957-39-1
:
Formula:
C₇H₄BrNO
MDL Number:
MFCD16628264
:
198.03 g/mol
Storage Temperature:
Ambient