Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dioxaspiro[4.5]decane-8-carbonitrile
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
69947-09-7
:
Formula:
C₉H₁₃NO₂
MDL Number:
MFCD09037879
:
167.2050 g/mol
Storage Temperature:
Ambient