Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Imidazo[4,5-b]pyridin-2(3H)-one
1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%
1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%
1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%
APOSOR953324-1G
:
Apollo Scientific
:
1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%
APOSOR953324-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
135.1200 g/mol
:
Ambient
:
16328-62-4
:
C₆H₅N₃O
:
MFCD00464845