Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-2,5-dimethoxy-4-nitrobenzene
1-Chloro-2,5-dimethoxy-4-nitrobenzene
1-Chloro-2,5-dimethoxy-4-nitrobenzene
Irritant
1-Chloro-2,5-dimethoxy-4-nitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Chloro-2,5-dimethoxy-4-nitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
34633-69-7
:
Formula:
C₈H₈ClNO₂
MDL Number:
MFCD00087057
:
185.610 g/mol
Melting Pt:
78…79 °C
:
GHS07
Storage Temperature:
Ambient