3-Methyl-3H-imidazo[4,5-b]pyridine 98%
Catalog # APOSOR952327-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.15 g/mol
Storage Temperature:Ambient
CAS Number:6688-61-5
Formula:C₇H₇N₃
MDL Number:MFCD03676333