Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-3,5-dinitrotoluene
4-Chloro-3,5-dinitrotoluene
4-Chloro-3,5-dinitrotoluene
Irritant
4-Chloro-3,5-dinitrotoluene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-3,5-dinitrotoluene
Supplier:
Apollo Scientific
About this item
CAS Number:
5264-65-3
:
Formula:
C₇H₅ClN₂O₄
MDL Number:
MFCD00060535
:
216.58 g/mol
Melting Pt:
114…116 °C
:
GHS07
Storage Temperature:
Ambient