Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methyl-1,8-naphthyridine 98%

2-Methyl-1,8-naphthyridine 98%

Supplier:   Apollo Scientific
2-Methyl-1,8-naphthyridine 98%
Supplier:  Apollo Scientific
Certificates

6 Options Available Below

Product Details & Documents
Synonyms:  2-Methyl-1,8-diazanaphthalene, 2-Methylpyrido[2,3-b]pyridine, 2-Methyl[1,8]-Naphthyridine
  • CAS Number:1569-16-0
  • Formula:C₉H₈N₂
  • MDL Number:MFCD04066717
  • Molecular Weight:144.18 g/mol
  • Storage Temperature:Ambient
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR951676-250MG
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR951676-1G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 30,40 €
    30,40 €
    1 * 5 g
       30,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR951676-5G
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 148,00 €
    148,00 €
    1 * 25 g
       148,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR951676-25G
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 585,00 €
    585,00 €
    1 * 100 g
       585,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR951676-100G
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2 920,00 €
    2 920,00 €
    1 * 500 g
       2 920,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR951676-500G
    New Product