Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Pyrazoledicarboxylic acid
3,5-Pyrazoledicarboxylic acid 95%
3,5-Pyrazoledicarboxylic acid 95%
3,5-Pyrazoledicarboxylic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Pyrazoledicarboxylic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
3112-31-0
:
Formula:
C₅H₄N₂O₄
MDL Number:
MFCD00005235
:
156.10 g/mol
Melting Pt:
292…295 °C
Storage Temperature:
Ambient