Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3,4-Dichlorophenyl)ethylamine
1-(3,4-Dichlorophenyl)ethylamine 97%
1-(3,4-Dichlorophenyl)ethylamine 97%
1-(3,4-Dichlorophenyl)ethylamine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(3,4-Dichlorophenyl)ethylamine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
74877-07-9
:
Formula:
C₈H₉Cl₂N
MDL Number:
MFCD01123279
:
190.07 g/mol
Storage Temperature:
Ambient