Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Isopropyl-N,2,3-trimethylbutyramide
2-Isopropyl-N,2,3-trimethylbutyramide 95%
2-Isopropyl-N,2,3-trimethylbutyramide 95%
2-Isopropyl-N,2,3-trimethylbutyramide 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Isopropyl-N,2,3-trimethylbutyramide 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
51115-67-4
:
Formula:
C₁₀H₂₁NO
MDL Number:
MFCD00130070
:
171.28 g/mol
Storage Temperature:
Ambient