Order Entry
Export
ContactUsLinkComponent
tert-Butoxytrimethylsilane 95%
tert-Butoxytrimethylsilane 95%
Catalog # APOSOR948280-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butoxytrimethylsilane 95%
Catalog # APOSOR948280-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :146.31 g/mol
Storage Temperature:Ambient
CAS Number:13058-24-7
Formula:C₇H₁₈OSi
MDL Number:MFCD00054893