Order Entry
Export
ContactUsLinkComponent
2,5-Dimethyl-4-methoxybenzoic acid 95%
2,5-Dimethyl-4-methoxybenzoic acid 95%
Catalog # APOSOR944980-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethyl-4-methoxybenzoic acid 95%
Catalog # APOSOR944980-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.21 g/mol
Storage Temperature:Ambient
CAS Number:58106-26-6
Formula:C₁₀H₁₂O₃
MDL Number:MFCD01194285