2,2-Bis(hydroxymethyl)propionic acid 98%
Catalog # APOSOR9441-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:134.13 g/mol
Melting Pt:189…191 °C
Storage Temperature:Ambient
CAS Number:4767-03-7
Formula:C₅H₁₀O₄
MDL Number:MFCD00004199