Order Entry
Export
ContactUsLinkComponent
 
N-Methyl-1,3-benzothiazol-2-amine 95%
Catalog # APOSOR943526-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-1,3-benzothiazol-2-amine 95%
Catalog # APOSOR943526-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.23 g/mol
Storage Temperature:Ambient
CAS Number:16954-69-1
Formula:C₈H₈N₂S
MDL Number:MFCD00035877