0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Methyl 2,4-dimethoxybenzoate
Methyl-2,4-dimethoxybenzoate 95%
Methyl-2,4-dimethoxybenzoate 95%
Methyl-2,4-dimethoxybenzoate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-2,4-dimethoxybenzoate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2150-41-6
:
Formula:
C₁₀H₁₂O₄
MDL Number:
MFCD00070649
:
196.2 g/mol
Storage Temperature:
Ambient