
2,3-Piperazinedione 95%
Catalog # APOSOR942215-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:114.1 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:13092-86-9
Formula:C₄H₆N₂O₂