Order Entry
Export
ContactUsLinkComponent
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 95%
Flammable
Irritant
Health
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 95%
Catalog # APOSOR939803-10G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Health
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 95%
Catalog # APOSOR939803-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :124.18 g/mol
Boiling Pt:159 °C
Melting Pt:–100 °C
Density:0.96 g/cm³ (25 °C)
Flash Pt:58 °C
UN:3271
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:106-86-5
Formula:C₈H₁₂O
MDL Number:MFCD00022356