Order Entry
Export
ContactUsLinkComponent
2,2-Dimethylhexanenitrile 95%
2,2-Dimethylhexanenitrile 95%
Catalog # APOSOR939269-5G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethylhexanenitrile 95%
Catalog # APOSOR939269-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :125.22 g/mol
Storage Temperature:Ambient
CAS Number:55897-65-9
Formula:C₈H₁₅N