Order Entry
Export
ContactUsLinkComponent
2,2-Dimethylvaleronitrile 95%
2,2-Dimethylvaleronitrile 95%
Catalog # APOSOR939158-5G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethylvaleronitrile 95%
Catalog # APOSOR939158-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :111.19 g/mol
Storage Temperature:Ambient
CAS Number:20654-47-1
Formula:C₇H₁₃N