Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Bis(phenylsulphinyl)ethane
1,2-Bis(phenylsulphinyl)ethane 97%
1,2-Bis(phenylsulphinyl)ethane 97%
:
Apollo Scientific
1,2-Bis(phenylsulphinyl)ethane 97%
:
Apollo Scientific
:
6099-21-4
:
:
C₁₄H₁₄O₂S₂
:
MFCD11846120
:
278.38 g/mol
:
GHS07
:
Ambient