Order Entry
Export
ContactUsLinkComponent
 
(2,4-Dichloro-3-methylphenyl)amine hydrochloride 95%
Catalog # APOSOR938922-1G
Supplier:  Apollo Scientific
CAS Number:  
(2,4-Dichloro-3-methylphenyl)amine hydrochloride 95%
Catalog # APOSOR938922-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.51 g/mol
Storage Temperature:Ambient
CAS Number:19853-79-3
Formula:C₇H₈Cl₃N
MDL Number:MFCD13186332