Order Entry
Export
ContactUsLinkComponent
1-Ethyl-1,4-diazepane 95%
1-Ethyl-1,4-diazepane 95%
Catalog # APOSOR938795-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Ethyl-1,4-diazepane 95%
Catalog # APOSOR938795-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :128.22 g/mol
Storage Temperature:Ambient
CAS Number:3619-73-6
Formula:C₇H₁₆N₂
MDL Number:MFCD06808650