Order Entry
Export
ContactUsLinkComponent
2,3-Thiophenedicarboxylic anhydride 95%
2,3-Thiophenedicarboxylic anhydride 95%
Catalog # APOSOR938656-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Thiophenedicarboxylic anhydride 95%
Catalog # APOSOR938656-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.14 g/mol
Storage Temperature:Ambient
CAS Number:6007-83-6
Formula:C₆H₂O₃S
MDL Number:MFCD09832453