Order Entry
Export
ContactUsLinkComponent
 
3-Bromo-2,2-dimethyl-1-propanol 95%
Catalog # APOSOR938619-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2,2-dimethyl-1-propanol 95%
Catalog # APOSOR938619-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :167.05 g/mol
Storage Temperature:Ambient
CAS Number:40894-00-6
Formula:C₅H₁₁BrO
MDL Number:MFCD00004683