Pyridine-2,6-dicarbonitrile 95%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,6-Dicyanopyridine
- CAS Number:2893-33-6
- Formula:C₇H₃N₃
- MDL Number:MFCD00129021
- Molecular Weight:129.12 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 2,6-Dicyanopyridine
- CAS Number:2893-33-6
- Formula:C₇H₃N₃
- MDL Number:MFCD00129021
- Molecular Weight:129.12 g/mol
- Storage Temperature:Ambient



