2,2-Diphenylethanol 98%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:1883-32-5
- Formula:C₁₄H₁₄O
- MDL Number:MFCD00004729
- Molecular Weight:198.27 g/mol
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
- CAS Number:1883-32-5
- Formula:C₁₄H₁₄O
- MDL Number:MFCD00004729
- Molecular Weight:198.27 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



