Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Dimethylcyclohexanone 95%

Supplier:   Apollo Scientific
3,4-Dimethylcyclohexanone 95%
Supplier:  Apollo Scientific
Certificates
3,4-Dimethylcyclohexanone 95%

3 Options Available Below

Product Details & Documents
  • CAS Number:5465-09-8
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00045552
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 225,00 €
    225,00 €
    1 * 1 g
       225,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 550,00 €
    550,00 €
    1 * 5 g
       550,00 €

  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 992,00 €
    992,00 €
    1 * 10 g
       992,00 €